Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npm1
Entrez Gene ID
394478
Gene Name
nucleophosmin (nucleolar phosphoprotein B23, numatrin)
Gene Aliases
b23, npm, numatrin
Chromosome Location
Chr.3: 9664012 - 9674013 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_203552.1 NP_988883.1 3 - 4 319 75
XM_012958741.2 XP_012814195.1 3 - 4 342 75
GenBank mRNA BC059736.1 - 3 - 4 319 75
CR761659.1 - 3 - 4 282 75

View Full Product Details