The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213692.1 | NP_998857.1 | 1 - 1 | 1554 | 82 |
GenBank mRNA | BC067916.1 | - | 1 - 1 | 1554 | 82 |
CR762327.2 | - | 1 - 1 | 1545 | 82 |