Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nme2
Entrez Gene ID
448725
Gene Name
NME/NM23 nucleoside diphosphate kinase 2
Gene Aliases
NM23-X1, ndpk-b, ndpkb, nm23-h2, nm23b, nm23ndk, nme2-a, nme2-b, puf
Chromosome Location
Chr.Un|NW_016683511: 179960 - 183736 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005140.2 NP_001005140.1 4 - 5 382 98
XM_012954135.1 XP_012809589.1 3 - 4 350 98
GenBank mRNA BC077684.1 - 4 - 5 383 98
CR760723.2 - 3 - 4 351 98

View Full Product Details