Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rpl12
Entrez Gene ID
493246
Gene Name
ribosomal protein L12
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683647: 45182 - 49316 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007859.1 NP_001007860.1 4 - 5 732 94
GenBank mRNA BC079961.1 - 4 - 5 732 94
CR761927.2 - 4 - 5 556 94

View Full Product Details