Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ints14
Entrez Gene ID
493315
Gene Name
integrator complex subunit 14
Gene Aliases
c15orf44, vwa9
Chromosome Location
Chr.3: 117437748 - 117451332 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007938.1 NP_001007939.1 11 - 12 1456 61
XM_012958881.2 XP_012814335.1 11 - 12 1890 61
GenBank mRNA BC080444.1 - 11 - 12 1456 61

View Full Product Details