Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cbwd2
Entrez Gene ID
496702
Gene Name
COBW domain containing 1, gene 2
Gene Aliases
cbwd1.2, cbwd6
Chromosome Location
Chr.1: 116757466 - 116789797 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011255.1 NP_001011255.1 7 - 8 552 106
XM_012961026.2 XP_012816480.1 8 - 9 671 106
XM_012961031.2 XP_012816485.1 8 - 9 566 106
GenBank mRNA BC086500.2 - 7 - 8 552 106

View Full Product Details