Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dctn5
Entrez Gene ID
496622
Gene Name
dynactin subunit 5
Gene Aliases
dctn4
Chromosome Location
Chr.9: 15525802 - 15531688 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011198.2 NP_001011198.1 3 - 4 253 64
GenBank mRNA BC087561.2 - 3 - 4 253 64

View Full Product Details