Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
crip3
Entrez Gene ID
548528
Gene Name
cysteine-rich protein 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683871: 722 - 6050 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015811.1 NP_001015811.1 2 - 3 436 64
GenBank mRNA BC090092.1 - 2 - 3 436 64
CR762212.2 - 2 - 3 350 64

View Full Product Details