Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
txnl4a
Entrez Gene ID
549400
Gene Name
thioredoxin like 4A
Gene Aliases
-
Chromosome Location
Chr.6: 28824037 - 28830646 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016646.2 NP_001016646.1 1 - 2 171 106
GenBank mRNA BC157181.1 - 2 - 3 243 106
CR760391.2 - 1 - 2 171 106

View Full Product Details