Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eif3g
Entrez Gene ID
549503
Gene Name
eukaryotic translation initiation factor 3 subunit G
Gene Aliases
eif3g-a, eif3g-b, eif3s4
Chromosome Location
Chr.3: 126567014 - 126577210 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016749.1 NP_001016749.1 9 - 10 951 90
GenBank mRNA BC158150.1 - 9 - 10 1095 90
CR855811.1 - 9 - 10 951 90

View Full Product Details