Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srd5a2
Entrez Gene ID
549867
Gene Name
steroid-5-alpha-reductase, alpha polypeptide 2 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 2)
Gene Aliases
SRD5alpha2
Chromosome Location
Chr.5: 40593253 - 40630460 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017113.2 NP_001017113.1 5 - 6 914 102
XM_018093906.1 XP_017949395.1 5 - 6 1160 101
GenBank mRNA CR761203.2 - 5 - 6 914 102

View Full Product Details