Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cldnd1
Entrez Gene ID
549013
Gene Name
claudin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 61867488 - 61902151 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016259.3 NP_001016259.1 3 - 4 670 79
XM_012968378.2 XP_012823832.1 4 - 5 802 79
XM_012968380.2 XP_012823834.1 3 - 4 577 79
XM_018095391.1 XP_017950880.1 4 - 5 843 79
GenBank mRNA CR761716.2 - 3 - 4 671 79

View Full Product Details