Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
gpbp1l1
Entrez Gene ID
549266
Gene Name
GC-rich promoter binding protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.4: 73370641 - 73411699 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016512.2 NP_001016512.1 6 - 7 875 66
XM_012961097.2 XP_012816551.1 7 - 8 1801 66
XM_012961098.2 XP_012816552.1 7 - 8 1799 66
XM_012961099.2 XP_012816553.1 8 - 9 1875 66
XM_012961100.2 XP_012816554.1 7 - 8 1795 66
XM_012961102.2 XP_012816556.1 7 - 8 1792 66
XM_018093050.1 XP_017948539.1 6 - 7 998 66
GenBank mRNA BC170710.1 - 5 - 6 828 66
BC170712.1 - 5 - 6 828 66
CR926227.2 - 6 - 7 875 66

View Full Product Details