Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
meig1
Entrez Gene ID
549276
Gene Name
meiosis/spermiogenesis associated 1
Gene Aliases
-
Chromosome Location
Chr.3: 71622103 - 71626002 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016522.2 NP_001016522.1 2 - 3 207 94
GenBank mRNA BC170626.1 - 2 - 3 170 94
BC170628.1 - 2 - 3 170 94
CR926206.2 - 2 - 3 207 94

View Full Product Details