Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
znhit3
Entrez Gene ID
733819
Gene Name
zinc finger, HIT-type containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 64552101 - 64557711 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045581.1 NP_001039046.1 1 - 2 116 80
GenBank mRNA BC121593.1 - 1 - 2 116 80
BC157154.1 - 1 - 2 150 80
CR848407.2 - 1 - 2 81 80

View Full Product Details