Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olig4
Entrez Gene ID
734024
Gene Name
oligodendrocyte transcription factor 4
Gene Aliases
olig3.2
Chromosome Location
Chr.2: 82203937 - 82204979 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045715.1 NP_001039180.1 1 - 1 542 71
GenBank mRNA BC161514.1 - 1 - 1 536 71
CR848409.2 - 1 - 1 542 71

View Full Product Details