The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078853.1 | NP_001072321.1 | 3 - 4 | 906 | 106 |
XM_012966732.1 | XP_012822186.1 | 3 - 4 | 494 | 106 | |
XM_018095430.1 | XP_017950919.1 | 3 - 4 | 480 | 106 | |
GenBank mRNA | BC121389.1 | - | 3 - 4 | 906 | 106 |