Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nacc2
Entrez Gene ID
100125188
Gene Name
NACC family member 2, BEN and BTB (POZ) domain containing
Gene Aliases
btbd14a
Chromosome Location
Chr.Un|NW_016683625: 38856 - 128811 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103074.1 NP_001096544.1 2 - 3 1440 94
XM_012967897.2 XP_012823351.1 2 - 3 989 94
XM_012967898.2 XP_012823352.1 2 - 3 1034 94
GenBank mRNA BC136033.1 - 2 - 3 1440 94

View Full Product Details