Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dach2
Entrez Gene ID
100101662
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.8: 41464804 - 41791220 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110045.1 NP_001103515.1 6 - 7 1214 74
XM_018096093.1 XP_017951582.1 7 - 8 1499 74
GenBank mRNA BC136082.1 - 6 - 7 1208 74
BC153333.1 - 6 - 7 1214 74

View Full Product Details