Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prickle2
Entrez Gene ID
100126208
Gene Name
prickle planar cell polarity protein 2
Gene Aliases
-
Chromosome Location
Chr.4: 132243973 - 132459709 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012961768.2 XP_012817222.1 3 - 4 904 104
XM_012961769.1 XP_012817223.1 3 - 4 546 104
XM_012961771.2 XP_012817225.1 3 - 4 381 104
GenBank mRNA BC153335.1 - 3 - 4 653 104

View Full Product Details