Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
golph3
Entrez Gene ID
733503
Gene Name
golgi phosphoprotein 3 (coat-protein)
Gene Aliases
-
Chromosome Location
Chr.1: 187523432 - 187534911 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114215.1 NP_001107687.1 1 - 2 312 79
GenBank mRNA BC158266.1 - 1 - 2 312 79
CR760594.2 - 1 - 2 363 79
CR942407.2 - 1 - 2 300 79
CT485738.1 - 1 - 2 366 79

View Full Product Details