Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrg3
Entrez Gene ID
100125039
Gene Name
neuregulin 3
Gene Aliases
-
Chromosome Location
Chr.7: 42972258 - 43341392 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114241.1 NP_001107713.1 5 - 6 1449 100
GenBank mRNA BC153324.1 - 5 - 6 1449 100

View Full Product Details