Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wsb2
Entrez Gene ID
100145242
Gene Name
WD repeat and SOCS box containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 156678210 - 156699447 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126726.1 NP_001120198.1 3 - 4 659 92
XM_018090799.1 XP_017946288.1 3 - 4 847 92
GenBank mRNA BC159319.1 - 3 - 4 659 92

View Full Product Details