Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wsb2
Entrez Gene ID
100145242
Gene Name
WD repeat and SOCS box containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 156678210 - 156699447 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126726.1 NP_001120198.1 4 - 5 902 90
GenBank mRNA BC159319.1 - 4 - 5 902 90

View Full Product Details