Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ubfd1
Entrez Gene ID
100145608
Gene Name
ubiquitin family domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 16732416 - 16743320 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127017.1 NP_001120489.1 6 - 7 865 96
XM_012970101.2 XP_012825555.1 6 - 7 801 96
GenBank mRNA BC161365.1 - 6 - 7 865 96

View Full Product Details