Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
spock2
Entrez Gene ID
100144943
Gene Name
SPARC (osteonectin), cwcv and kazal like domains proteoglycan 2
Gene Aliases
-
Chromosome Location
Chr.7: 34724180 - 34819993 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126515.1 NP_001119987.1 7 - 8 969 78
XM_012966046.2 XP_012821500.1 8 - 9 1101 78
GenBank mRNA BC158244.1 - 7 - 8 969 78

View Full Product Details