Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrn4
Entrez Gene ID
100145335
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
-
Chromosome Location
Chr.5: 21087075 - 21096557 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126807.1 NP_001120279.1 3 - 4 1011 102
GenBank mRNA BC160543.1 - 3 - 4 1011 102

View Full Product Details