Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxo47
Entrez Gene ID
100158537
Gene Name
F-box protein 47
Gene Aliases
-
Chromosome Location
Chr.10: 3643075 - 3667151 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127973.2 NP_001121445.2 9 - 10 1145 73
GenBank mRNA BC166198.1 - - 1134 73

View Full Product Details