Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
abhd2
Entrez Gene ID
100216078
Gene Name
abhydrolase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 99654934 - 99691049 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142063.1 NP_001135535.1 6 - 7 838 91
XM_018091965.1 XP_017947454.1 6 - 7 912 91
GenBank mRNA BC168004.1 - 6 - 7 838 91

View Full Product Details