Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spry1
Entrez Gene ID
449470
Gene Name
sprouty RTK signaling antagonist 1
Gene Aliases
Xtsprouty1, hspry1, sprouty, sprouty-1, sprouty1, spry1-a, spry1-b, xspry1
Chromosome Location
Chr.1: 56196385 - 56201486 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006930.2 NP_001006931.2 2 - 2 1349 101
XM_012960230.2 XP_012815684.1 2 - 2 1456 101
GenBank mRNA AY714336.2 - 1 - 1 808 101
BC135278.1 - 2 - 2 1349 101

View Full Product Details