Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
acs7
Entrez Gene ID
100279981
Gene Name
1-aminocyclopropane-1-carboxylate synthase7
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001152929.1 NP_001146401.1 - 628 56
GenBank mRNA BT055116.1 - - 628 56
BT055282.1 - - 862 56

View Full Product Details