Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
csy1
Entrez Gene ID
542482
Gene Name
Chloroplast SecY-1
Gene Aliases
SECY
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112035.2 NP_001105505.2 - 1288 107
GenBank mRNA AF039304.1 - - 1308 107
BT039711.1 - - 1288 107

View Full Product Details