Gene Transcripts
- Gene Symbol
- Cdk3-ps
- Entrez Gene ID
- 69681
- Gene Name
- cyclin-dependent kinase 3, pseudogene
- Gene Aliases
- 2310050O10Rik, Cdk3
- Chromosome Location
- Chr. 11: 116216002 - 116220287 on Build GRCm38
- UniGene ID
- Mm.33677
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004853.1 |
|
7 |
1279 |
|
GenBank mRNA |
AK009918.1 |
- |
5 |
1105 |
|
AK166387.1 |
- |
2 |
592 |
|
View Full Product Details