Gene Transcripts
- Gene Symbol
- Sprr2j-ps
- Entrez Gene ID
- 20764
- Gene Name
- small proline-rich protein 2J, pseudogene
- Gene Aliases
- Sprr2j
- Chromosome Location
- Chr. 3: 92418087 - 92419396 on Build GRCm38
- UniGene ID
- Mm.57084
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003185.1 |
|
2 |
615 |
|
GenBank mRNA |
AK019074.1 |
- |
2 |
617 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY708877.1 |
- |
2 |
617 |
|
Chr. 3 - 92418087 - 92419398 [+] |
View Full Product Details