Gene Transcripts
- Gene Symbol
- Otx2os1
- Entrez Gene ID
- 606497
- Gene Name
- orthodenticle homolog 2 opposite strand 1
- Gene Aliases
- Chromosome Location
- Chr. 14: 48669288 - 48793348 on Build GRCm38
- UniGene ID
- Mm.165199
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029384.1 |
|
|
3, 4 |
365 |
|
GenBank mRNA |
AK042665.1 |
- |
LIT3409 Otx2OS |
3, 4 |
365 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB246370.2 |
- |
3, 4 |
327 |
|
Chr. 14 - 48669326 - 48793345 [+] |
GenBankmRNA |
BB639822.1 |
- |
3, 4 |
365 |
|
Chr. 14 - 48669288 - 48793291 [+] |
GenBankmRNA |
BU504680.1 |
- |
2, 3 |
391 |
|
Chr. 14 - 48670159 - 48719799 [+] |
View Full Product Details