Gene Transcripts

Gene Symbol
AOX3P-AOX2P
Entrez Gene ID
107161151
Gene Name
AOX3P-AOX2P readthrough
Gene Aliases
AOX3/AOX3L1
Chromosome Location
Chr. 2: 200695723 - 200794915 on Build GRCh38
UniGene ID
Hs.148269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135011.1 31 4247
NR_135012.1 30 3740
GenBank mRNA JX431285.1 - 30 3547
JX431286.1 - 31 4054

View Full Product Details