Gene Transcripts
- Gene Symbol
- CSN1S2AP
- Entrez Gene ID
- 286828
- Gene Name
- casein alpha s2-like A, pseudogene
- Gene Aliases
- CSN1S2A
- Chromosome Location
- Chr. 4: 70067386 - 70085273 on Build GRCh38
- UniGene ID
- Hs.631945
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003720.1 |
|
13 |
635 |
|
GenBank mRNA |
AY154892.1 |
- |
13 |
635 |
|
View Full Product Details