Gene Transcripts
- Gene Symbol
- COL6A4P1
- Entrez Gene ID
- 344875
- Gene Name
- collagen type VI alpha 4 pseudogene 1
- Gene Aliases
- COL6A4, COL6A4P, DIVA, DVWA, VWA6
- Chromosome Location
- Chr. 3: 15165362 - 15205959 on Build GRCh38
- UniGene ID
- Hs.722756
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027927.1 |
|
2 |
666 |
|
GenBank mRNA |
AB299979.1 |
- |
2 |
666 |
|
FN394065.1 |
- |
2 |
500 |
|
View Full Product Details