Gene Transcripts
- Gene Symbol
- UBE2DNL
- Entrez Gene ID
- 100131816
- Gene Name
- ubiquitin conjugating enzyme E2 D N-terminal like (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. X: 84934151 - 84934890 on Build GRCh38
- UniGene ID
- Hs.460809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024062.1 |
|
1 |
642 |
|
GenBank mRNA |
BC040290.1 |
- |
1 |
642 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI341772.1 |
- |
2 |
394 |
|
Chr. X - 84934260 - 84934879 [+] |
GenBankmRNA |
DB527789.1 |
- |
1 |
376 |
|
Chr. X - 84934417 - 84934884 [+] |
View Full Product Details