Gene Transcripts
- Gene Symbol
- TTTY11
- Entrez Gene ID
- 83866
- Gene Name
- testis-specific transcript, Y-linked 11 (non-protein coding)
- Gene Aliases
- NCRNA00134, TTY11
- Chromosome Location
- Chr. Y: 8783313 - 8817382 on Build GRCh38
- UniGene ID
- Hs.122396
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001548.2 |
|
2 |
107 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF332240.3 |
- |
2 |
107 |
|
Chr. Y - 8783310 - 8817382 [-] |
View Full Product Details