Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_028052.1 | 1 | 331 | ||
GenBank mRNA | AB029315.1 | - | 1 | 330 | |
AF242770.1 | - | 1 | 286 | ||
AK312641.1 | - | 2 | 135 | ||
AL355700.1 | - | 1 | 282 | ||
AL360142.1 | - | 1 | 303 | ||
AL834503.1 | - | 1 | 321 | ||
BC007222.1 | - | 1 | 306 | 2819984 | |
BC009389.2 | - | 1 | 281 | 4130266 | |
BC017192.2 | - | 1 | 287 | 3139180 | |
BC019023.2 | - | 1 | 287 |