Gene Transcripts
- Gene Symbol
- SCARNA7
- Entrez Gene ID
- 677767
- Gene Name
- small Cajal body-specific RNA 7
- Gene Aliases
- U90
- Chromosome Location
- Chr. 3: 160514907 - 160515236 on Build GRCh38
- UniGene ID
- Hs.675953
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003001.1 |
|
1 |
118 |
|
GenBank mRNA |
AY077740.1 |
- |
1 |
118 |
|
View Full Product Details