Gene Transcripts
- Gene Symbol
- SERPINA13P
- Entrez Gene ID
- 388007
- Gene Name
- serpin family A member 13, pseudogene
- Gene Aliases
- SERPINA13, UNQ6121
- Chromosome Location
- Chr. 14: 94640725 - 94646994 on Build GRCh38
- UniGene ID
- Hs.527795
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015340.1 |
|
1 |
62 |
|
GenBank mRNA |
AY358238.1 |
- |
1 |
62 |
|
View Full Product Details