Gene Transcripts
- Gene Symbol
- SNHG12
- Entrez Gene ID
- 85028
- Gene Name
- small nucleolar RNA host gene 12
- Gene Aliases
- ASLNC04080, C1orf79, LINC00100, NCRNA00100
- Chromosome Location
- Chr. 1: 28578538 - 28581854 on Build GRCh38
- UniGene ID
- Hs.632377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024127.1 |
|
5, 6 |
597 |
|
GenBank mRNA |
AF277181.1 |
- |
5, 6 |
385 |
|
AK092096.1 |
- |
2, 3 |
1766 |
|
AY277594.1 |
- |
3, 4 |
165 |
|
BC022205.1 |
- |
5, 6 |
1226 |
|
BC034611.1 |
- |
3, 4 |
1448 |
|
BC045792.1 |
- |
4, 5 |
956 |
|
BC064589.1 |
- |
3, 4 |
1487 |
|
BC068048.1 |
- |
5, 6 |
597 |
|
BC107730.1 |
- |
2, 3 |
989 |
|
KJ782215.1 |
- |
6, 7 |
1726 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN260333.1 |
- |
5, 6 |
602 |
|
Chr. 1 - 28578597 - 28581859 [-] |
GenBankmRNA |
BE794361.1 |
- |
5, 6 |
577 |
|
Chr. 1 - 28578538 - 28581834 [-] |
GenBankmRNA |
BF765335.1 |
- |
4, 5 |
287 |
|
Chr. 1 - 28578636 - 28581171 [-] |
GenBankmRNA |
AV697366.1 |
- |
|
546 |
|
Chr. 1 - 28579532 - 28581040 [-] |
GenBankmRNA |
EB387922.1 |
- |
3, 4 |
219 |
|
Chr. 1 - 28578536 - 28580649 [-] |
View Full Product Details