Gene Transcripts
- Gene Symbol
- UBE2Q2P1
- Entrez Gene ID
- 388165
- Gene Name
- ubiquitin conjugating enzyme E2 Q2 pseudogene 1
- Gene Aliases
- UBE2QP1
- Chromosome Location
- Chr. 15: 84527196 - 84570795 on Build GRCh38
- UniGene ID
- Hs.498348
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003661.2 |
|
4 |
1542 |
|
GenBank mRNA |
AK125266.1 |
- |
4 |
1542 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN414393.1 |
- |
5 |
544 |
|
Chr. 15 - 84542144 - 84570712 [-] |
View Full Product Details