Gene Transcripts
- Gene Symbol
- IFITM4P
- Entrez Gene ID
- 340198
- Gene Name
- interferon induced transmembrane protein 4 pseudogene
- Gene Aliases
- dJ377H14.5
- Chromosome Location
- Chr. 6: 29750807 - 29751148 on Build GRCh38
- UniGene ID
- Hs.450189
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001590.1 |
|
1 |
332 |
|
View Full Product Details