Gene Transcripts
- Gene Symbol
- SNORA54
- Entrez Gene ID
- 677833
- Gene Name
- small nucleolar RNA, H/ACA box 54
- Gene Aliases
- ACA54
- Chromosome Location
- Chr. 11: 2963771 - 2963893 on Build GRCh38
- UniGene ID
- Hs.689696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002982.1 |
|
1 |
39 |
|
GenBank mRNA |
AJ609479.1 |
- |
1 |
39 |
|
View Full Product Details