Gene Transcripts
- Gene Symbol
- HCG4B
- Entrez Gene ID
- 80868
- Gene Name
- HLA complex group 4B (non-protein coding)
- Gene Aliases
- HCG4P6, HCGIV-06, HCGIV-6, HCGIV.5, bCX67J3.3, bPG309N1.1, bQB90C11.3
- Chromosome Location
- Chr. 6: 29924592 - 29927215 on Build GRCh38
- UniGene ID
- Hs.661198
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001317.3 |
|
1 |
1677 |
|
GenBank mRNA |
AK056727.1 |
- |
2 |
1638 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF036973.1 |
- |
2 |
504 |
|
Chr. 6 - 29924695 - 29926086 [-] |
View Full Product Details