Gene Transcripts

Gene Symbol
Mkrn1-ps1
Entrez Gene ID
353327
Gene Name
makorin, ring finger protein 1, pseudogene 1
Gene Aliases
Chromosome Location
Chr. 5: 88837799 - 88839399 on Build GRCm38
UniGene ID
Mm.423672
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF494488.1 - LIT1345
Makorin1-p1
1 237

View Full Product Details