Gene Transcripts
- Gene Symbol
- SIAH1
- Entrez Gene ID
- 6477
- Gene Name
- siah E3 ubiquitin protein ligase 1
- Gene Aliases
- SIAH1A
- Chromosome Location
- Chr. 16: 48360531 - 48448440 on Build GRCh38
- UniGene ID
- Hs.706828
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023458.1 |
- |
1 |
1140 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA096768.1 |
- |
1 |
305 |
|
Chr. 16 - 48364477 - 48365050 [-] |
View Full Product Details